Learn More
Cayman Chemical AmInoacetonhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1757310 MG
Encompass_Preferred
A threonine and glycine catabolite that can be converted to methylglyoxal by amine oxidases; at 0.10-5 mM acts as a pro-oxidant inducing cell death in RINm5f pancreatic β-cells; used as a growth substrate for Pseudomonas
Catalog No. 50-296-5296