Learn More
Medchemexpress LLC Aminoacylase (Acylase I) | 9012-37-7 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYP296050MG
Encompass_Preferred
Aminoacylase (Acylase I) is a specific N-acetylglutamine metabolic-degrading enzyme. It serves as a key enzyme in the intracellular amino acid recycling pathway and offers detoxifying properties.
  • Solid appearance, off-white to light brown
  • Derived from porcine kidney
  • Activity of 1825 unit/mg protein
  • One unit hydrolyzes 1.0 micromole of N-Acetyl-L Methionine per hour at pH 7.0 at 25°C
Catalog No. 50-003-98329