Learn More
Medchemexpress LLC γ-Aminobutyric acid (Standard) | 56-12-2 | 99.9% | 103.12 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0067R50MG
Encompass_Preferred
γ-Aminobutyric acid (Standard) is the analytical standard for γ-Aminobutyric acid, intended for research and analytical applications. It is a major inhibitory neurotransmitter in the adult mammalian brain, acting by binding to ionotropic GABA receptors (GABAA receptors) and metabotropic receptors (GABAB receptors). This compound exhibits a calming effect by blocking specific signals within the central nervous system.
  • Analytical standard and reference standard used in assays.
  • Intended for research and analytical applications.
  • Frequently utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Functions as a major inhibitory neurotransmitter in the adult mammalian brain.
  • Shows a calming effect by blocking specific central nervous system signals.
  • Classified as an endogenous metabolite.
Catalog No. 50-004-27893