Learn More
Medchemexpress LLC Aminocaproyl-Val-Cit-PABC-MMAE | 1374407-35-8 | 99.9% | 1236.58 | 25 MG

Supplier: Medchemexpress LLC 25MGHY19697
Aminocaproyl-Val-Cit-PABC-MMAE is a drug-linker conjugate specifically designed for Antibody-Drug Conjugates (ADCs). It is intended for research use only and is not sold to patients. The product is available in solid form, appearing as a white to off-white powder with a purity of 99.89%. It is categorized under "Antibody-drug Conjugate/ADC Related" and "Drug-Linker Conjugates for ADC" within product classifications.
- High purity: 99.89%
- Specific application: drug-linker conjugate for ADC research
- Available documentation: data sheet, COA, SDS, handling instructions
- Storage recommendations: -20°C, sealed storage, away from moisture; unstable in solutions, freshly prepared is recommended.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.