Learn More
Cayman Chemical Aminoethoxyvinyl Glycine hyd
SDP
Supplier:  Cayman Chemical 1554625 MG
Encompass_Preferred
An ethylene biosynthesis inhibitor and a plant growth regulator; inhibits ethylene production in, and elongation of, maize root tips at 1 μM; inhibits cystathionine β-lyase and cystathionine γ-lyase (Kis = 1.1 and 10.5 μM, respectively); delays color change from green to red and maintains fruit firmness, markers of fruit ripening, in postharvest tomatoes at 1 mM
Catalog No. 50-296-9342