Learn More
Medchemexpress LLC Aminoethyl-SS-propionic acid | 15579-00-7 | 98.0% | 181.28 | C5H11NO2S2 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1400965G
Encompass_Preferred
Aminoethyl-SS-propionic acid is a cleavable disulfide-containing ADC linker used in the synthesis of antibody-drug conjugates (ADCs) for research. It provides a reducible linkage intended to release cytotoxic payloads in intracellular reducing environments. Typical properties include CAS 15579-00-7, formula C5H11NO2S2, and molecular weight 181.28.
  • Cleavable disulfide linker suitable for ADC construction.
  • Releases payloads under intracellular reducing conditions.
  • High purity for research applications.
  • Stable as a powder at -20°C for long-term storage.
  • Available in gram-scale quantities for synthesis scale-up.
Catalog No. 50-002-04163