Learn More
TOKU-E Puromycin Dihydrochloride, 58-58-2, 250 mg
Supplier:  TOKU-E P001250MG
Encompass_Preferred
aminoglycoside antibiotic, C22H29N7O5 2HCl, selection antibiotic, Inhibits protein synthesis. Useful in cell culture, cancer research, transfection.
Catalog No. 50-255-1770