Learn More
TOKU-E Puromycin Dihydrochloride, 58-58-2, 500 mg
Supplier:  TOKU-E P001500MG
Encompass_Preferred
aminoglycoside antibiotic, C22H29N7O5 2HCl, selection antibiotic, Inhibits protein synthesis. Useful in cell culture, cancer research, transfection.
Catalog No. 50-255-1769