Learn More
TOKU-E Hygromycin B, 31282-04-9, 1 g
Supplier:  TOKU-E H0241G
Encompass_Preferred
aminoglycoside antibiotic, selection antibiotic, C20H37N3O13, EvoPure, 99.0%, endotoxin ≤10 EU/mg. Useful in microbiology,, cell culture, plant biology.
Catalog No. 50-255-1787