Learn More
TOKU-E Amphotericin B, Solubilized, Lyophilized, 1397-89-3, 500 mg
Supplier:  TOKU-E A097500MG
Encompass_Preferred
aminoglycoside antimycotic, C47H73NO17. Useful in microbiology, viral transport media, cell culture, plant biology. Formulated with sodium deoxycholate.
Catalog No. 50-255-1837