Learn More
Medchemexpress LLC Aminoiminomethanesulphinic acid | 1758-73-2 | MFCD00002397 | 99.1% | 108.12 g/mol | CH4N2O2S | 1000 G
SDP
Supplier:  Medchemexpress LLC HYW0159151000G
Encompass_Preferred
Aminoiminomethanesulphinic acid (thiourea dioxide) is an organosulfur compound supplied for research and analytical use. It functions as a reducing agent and reagent in biochemical assays and industrial processes. The white to off-white solid has CAS 1758-73-2, molecular formula CH4N2O2S, and a molecular weight of 108.12 g/mol.
  • White to off-white solid, suitable for laboratory use.
  • High assay purity (≈99.06%).
  • Acts as a reducing agent in chemical and biochemical applications.
  • Soluble in water at ambient temperature.
  • Available in multiple pack sizes for research and scale-up.
Catalog No. 50-003-00578