Learn More
Medchemexpress LLC Aminoiminomethanesulphinic acid | 1758-73-2 | MFCD00002397 | 99.1% | 108.12 g/mol | CH4N2O2S | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW015915
Encompass_Preferred
Aminoiminomethanesulphinic acid (thiourea dioxide) is a small-molecule reducing agent used in chemical and life-science research. It is a white to off-white crystalline solid with molecular weight 108.12 g/mol and formula CH4N2O2S. Laboratory formats include solid and DMSO solutions for synthesis and reductive applications.
  • High purity: 99.06%.
  • Soluble in DMSO: 10 mg/mL (92.49 mM) with ultrasonic warming.
  • Available as solids and ready-to-use DMSO solutions.
  • Suitable for reductive degradation and synthesis reactions.
  • Use newly opened DMSO due to hygroscopic effects.
Catalog No. 50-003-03436