Learn More
Medchemexpress LLC Aminooxy-PEG2-alcohol | 185022-12-2 | MFCD30182047 | 121.14 | C4H11NO3 | 100 MG

Supplier: Medchemexpress LLC HY126951100MG
Aminooxy-PEG2-alcohol is a short PEG-based linker containing an aminooxy functional group and a terminal hydroxyl. It is used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based linker in PROTAC synthesis.
- Non-cleavable PEG linker for ADC and PROTAC synthesis.
- Contains an aminooxy group for selective oxime ligation and bioconjugation.
- Molecular formula C4H11NO3; molecular weight 121.14.
- Available pack sizes: 100 MG, 250 MG, 1 G.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.