Learn More
Medchemexpress LLC Aminooxy-PEG2-alcohol | 185022-12-2 | MFCD30182047 | 95.0% | 121.14 g·mol⁻¹ | C4H11NO3 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY126951250MG
Encompass_Preferred
Aminooxy-PEG2-alcohol is a short, bifunctional polyethylene glycol (PEG) linker bearing an aminooxy functional group and a terminal alcohol. It is used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based linker for PROTAC and other bioconjugation applications. Lot-specific purity and handling details are provided in the certificate of analysis.
Catalog No. 50-002-03166