Learn More
Medchemexpress LLC Aminooxy-PEG4-C2-Boc | 2100306-82-7 | >98% | C15H31NO7 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140402100MG
Encompass_Preferred
Aminooxy-PEG4-C2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • Used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker.
  • One ligand is for an E3 ubiquitin ligase.
  • The other ligand is for the target protein.
  • Exploits the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
Catalog No. 50-003-56897