Learn More
Medchemexpress LLC Aminooxy-PEG5-azide | 1919045-02-5 | 98.3% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13050750MG
Encompass_Preferred
Aminooxy-PEG5-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its azide group, allowing it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group for chemical reactions
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-38618