Learn More
Medchemexpress LLC Aminooxyacetic acid | 645-88-5 | MFCD01671589 | 95.0% | 91.07 g/mol | C2H5NO3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY107994A5MG
Encompass_Preferred
Aminooxyacetic acid is a research reagent that inhibits the malate-aspartate shuttle and the GABA-degrading enzyme GABA-T, and is used to probe cellular metabolism and neurotransmitter regulation in vitro and in vivo.
  • Inhibits the malate-aspartate shuttle (MAS) and GABA-T.
  • Useful for studies of cellular metabolism and neurotransmitter regulation.
  • Provided at 95.0% purity for research applications.
  • Molecular weight 91.07 g/mol and formula C2H5NO3.
  • Available in small research sizes, including 5 mg.
  • Includes downloadable COA and SDS for quality and safety information.
Catalog No. 50-002-42858