Learn More
Medchemexpress LLC Aminophenylthiazole | 552309-42-9 | 99.0% | 416.52 g·mol⁻¹ | C19H20N4O3S2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10061250MG
Encompass_Preferred
T16Ainh-A01 is an aminophenylthiazole small-molecule inhibitor of the calcium-activated chloride channel TMEM16A (ANO1). It inhibits TMEM16A-mediated chloride currents with an IC50 of approximately 1 μM and is supplied as a white to off-white solid for research use. Solubility: DMSO 83.33 mg/mL; DMF ≥10 mg/mL. Storage: powder -20°C (up to 3 years); in solvent -80°C (6 months), -20°C (1 month).
  • Inhibits TMEM16A with IC50 ≈ 1 μM.
  • High purity (99.04%) for reproducible results.
  • White to off-white solid suitable for analytical and biological assays.
  • High solubility in DMSO and good solubility in DMF.
  • Long-term powder stability at -20°C; defined solvent storage durations.
Catalog No. 50-002-13788