Learn More
Cayman Chemical AmInophylIn 500g
SDP
Supplier:  Cayman Chemical 22235500 G
Encompass_Preferred
A phosphodiesterase inhibitor (IC50 = 120 µM) and adenosine receptor antagonist; a complex of theophylline and ethylenediamine; has in vivo bronchodilator and vasodilator activity; suppresses visceral hypersensitivity to colorectal distension in rat IBS-D model; provides renoprotection against murine renal ischemia-reperfusion injury
Catalog No. 50-298-6173