Learn More
Cayman Chemical AmInopterIn 10mg
SDP
Supplier:  Cayman Chemical 2180210 MG
Encompass_Preferred
A synthetic folic acid derivative with anticancer and immunosuppressive properties; used in HAT media for selection in mammalian cell culture
Catalog No. 50-298-5846