Learn More
Cayman Chemical AmInopterIn 50mg
Supplier: Cayman Chemical 2180250 MG
A synthetic folic acid derivative with anticancer and immunosuppressive properties; used in HAT media for selection in mammalian cell culture