Learn More
Medchemexpress LLC HY-14518 5mg Medchemexpress, Aminopterin CAS:54-62-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY145185MG
Encompass_Preferred
Medchemexpress, HY-14518 5mg Aminopterin CAS:54-62-6 Aminopterin (4-Aminofolic acid), the 4-amino derivative of folic acid, is a folic acid antagonist. Aminopterin catalyses the reduction of folic acid to tetrahydrofolic acid, and competitively inhibits dihydrofolate reductase (DHFR) with a Ki of 3.7 pM. Aminopterin has anticancer and immunosuppressive activity. Aminopterin is used in treatment of pediatric leukemia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9907