Learn More
Apexbio Technology LLC Aminothiazole 96-50-4 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B16595.1
Encompass_Preferred
Aminothiazole (96-50-4) is a small-molecule inhibitor targeting thyroid peroxidase It is designed to interfere with thyroid hormone biosynthesis thereby regulating thyroid hormone levels Aminothiazole exerts its biological activity primarily through inhibition of the thyroid peroxidase enzyme In in vitro studies aminothiazole demonstrates inhibitory activity with an IC50 value typically ranging from 1 to 10 M Based on these pharmacological properties aminothiazole holds research potential in the investigation of thyroid hormone regulatory mechanisms and pathologies such as hyperthyroidism as well as in antimicrobial screening assays
Catalog No. 50-000-89831