Learn More
Medchemexpress LLC Aminoxyacetic acid | 645-88-5 | 95.0% | 91.07 | C2H5NO3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY107994A1MG
Encompass_Preferred
Aminoxyacetic acid is a small-molecule research reagent that inhibits the malate-aspartate shuttle (MAS) and the GABA-degrading enzyme GABA-T. It is supplied in small laboratory pack sizes and is documented with analytical certificates to support research use.
  • Inhibits the malate-aspartate shuttle (MAS).
  • Inhibits GABA-transaminase (GABA-T).
  • Supplied in small research pack sizes for laboratory use.
  • Reported purity 95.0% for experimental consistency.
  • Documentation includes COA and SDS for quality verification.
  • Suitable for biochemical and pharmacological assays.
Catalog No. 50-002-42857