Learn More
Cayman Chemical AmInoxyACtIc ACDhydrochl 250mg
SDP
Supplier:  Cayman Chemical 28298250 MG
Encompass_Preferred
A GABA-T inhibitor (Ki = 9.16 μM) that induces GABA accumulation in the brain; inhibits CBS and CSE (IC50s = 8.5 and 1.1 μM, respectively); decreases GABA-T activity and increases GABA levels in all regions of rat brain at 100 mg/kg; decreases GABA-T activity and increases GABA in mouse brain at 13 mg/kg; inhibits 3-mercaptopropionic acid-, strychnine-, and pentetrazole-induced seizures in mice (ED50s = 53, 130, and 85 mg/kg, respectively); exhibits neurotoxicity in the chimney test (ED50 = 62 mg/kg) and is lethal (LD50 = 105 mg/kg) to mice
Catalog No. 50-294-0416