Learn More
Medchemexpress LLC Amiodarone-d10 hydrochloride | 1261393-77-4 | 98.8% | C25H20D10ClI2NO3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY14187S1MG
Encompass_Preferred
Amiodarone-d10 hydrochloride is a deuterium-labeled form of Amiodarone, an antiarrhythmic agent known for inhibiting ATP-sensitive potassium channels. It can serve as a tracer and internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. Deuteration of drug molecules is being studied for its potential to impact pharmacokinetic and metabolic profiles.
  • Deuterium labeled antiarrhythmic agent
  • Inhibits ATP-sensitive potassium channel
  • Used as a tracer and internal standard for quantitative analysis
  • Potential to influence pharmacokinetic and metabolic profiles
Catalog No. 50-003-54358