Learn More
Medchemexpress LLC Amiodarone hydrochloride | 19702-44-2 | MFCD00079083 | C25H30ClI2NO3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14188R100MG
Encompass_Preferred
Amiodarone hydrochloride is a class III antiarrhythmic agent used in the treatment of various ventricular and supraventricular arrhythmias. It works by prolonging the action potential and refractory period in myocardial tissue.
  • Acts as an antagonist at α1, β1, and β2 adrenoceptors
  • Functions as a non-competitive antagonist at muscarinic receptors
  • Blocks cardiac K+ channels and L-type Ca2+ channels
  • Displays anti-anginal, anti-fibrillatory, and vasodilator effects
Catalog No. 50-003-53440