Learn More
Cayman Chemical AmIodaronhydrochlorIde 10g
SDP
Supplier:  Cayman Chemical 1521310 G
Encompass_Preferred
A class III antiarrhythmic agent; inhibits KCNH2 with an IC50 value of 1 µM; also inhibits CYP2C8 and CYP3A4 in vitro at low micromolar concentrations
Catalog No. 50-296-9602