Learn More
TARGETMOL CHEMICALS INC Amiselimod hydrochloride 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1030510MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Amiselimod hydrochloride (MT-1303 hydrochloride) is a sphingosine 1-phosphate receptor-1 (S1P1) modulator. Purity 99.68%
Catalog No. 50-287-7670