Learn More
Medchemexpress LLC Amisulpride hydrochloride | 81342-13-4 | 99.79% | C17H28ClN3O4S | 1 ML
Supplier: Medchemexpress LLC HY14545A10MM/1ML
Amisulpride hydrochloride is a potent dopamine D2/D3 receptor antagonist, demonstrating Ki values of 2.8 nM and 3.2 nM for human dopamine D2 and D3 receptors, respectively. This compound is known to inhibit quinpirole-elicited [3H]thymidine incorporation and subtly enhance [3H]dopamine release from rat striatum slices. It is primarily intended for research applications.
- Dopamine D2/D3 receptor antagonist
- Targets human dopamine D2 and D3 receptors
- Inhibits quinpirole-elicited [3H]thymidine incorporation
- Increases [3H]dopamine release
- Involved in GPCR/G Protein pathway
- Involved in Neuronal Signaling pathway