Learn More
Medchemexpress LLC Amisulpride (hydrochloride) | 81342-13-4 | MFCD09836932 | 99.8% | C17H28ClN3O4S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14545A50MG
Encompass_Preferred
Amisulpride hydrochloride is a dopamine D2/D3 receptor antagonist with Kis of 2.8 and 3.2 nM for human dopamine D2 and D3, respectively. It is intended for research use only.
  • Dopamine D2/D3 receptor antagonist
  • Ki values of 2.8 nM for human dopamine D2 and 3.2 nM for human dopamine D3
  • Intended for research use only
  • Stable under recommended storage conditions
  • Appearance is a colorless to light yellow oil
Catalog No. 50-003-53998