Learn More
Cayman Chemical AmIsulprIde N-oxIde 25mg
SDP
Supplier:  Cayman Chemical 3544725 MG
Encompass_Preferred
A degradation product of amisulpride; formed via oxidation by ozonation or photodegradation; has been found as a surface water contaminant
Catalog No. 50-291-3932