Learn More
Medchemexpress LLC Amitifadine (hydrochloride) | 410074-74-7 | 99.6% | 264.58 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY18332A10MM/1ML
Encompass_Preferred
Amitifadine hydrochloride is a serotonin-norepinephrine-dopamine reuptake inhibitor (SNDRI). It exhibits IC50 values of 12 nM for serotonin, 23 nM for norepinephrine, and 96 nM for dopamine in HEK 293 cells. It is for research use only and not sold to patients.
  • Serotonin: 12 nM
  • Norepinephrine: 23 nM
  • Dopamine: 96 nM
  • Inhibition of [3H]dopamine uptake: 159 nM
  • Inhibition of [3H]serotonin uptake: 411 nM
  • Inhibition of [3H]noradrenaline uptake: 71 nM
Catalog No. 50-003-87541