Learn More
Medchemexpress LLC Amitifadine hydrochloride | 410074-74-7 | 99.6% | 264.58 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY18332A50MG
Encompass_Preferred
Amitifadine hydrochloride is a serotonin-norepinephrine-dopamine reuptake inhibitor (SNDRI). It exhibits IC50 values of 12 nM for serotonin, 23 nM for norepinephrine, and 96 nM for dopamine in HEK 293 cells.
  • Functions as a triple monoamine reuptake inhibitor, affecting serotonin, norepinephrine, and dopamine reuptake.
  • High purity of 99.64%.
  • Soluble in DMSO at concentrations ≥ 100 mg/mL and in H2O at 50 mg/mL.
  • Studied in clinical trials for major depressive disorder.
  • Strictly for research purposes.
Catalog No. 50-003-87542