Learn More
Medchemexpress LLC Amitraz-d3 | 33089-61-1 | 99.5% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1111S15MG
Encompass_Preferred
Amitraz-d3 is the deuterium labeled Amitraz. Amitraz is a non-systemic acaricide and insecticide, with alpha-adrenergic agonist activity, interaction with octopamine receptors of the central nervous system and inhibition of monoamine oxidases and prostaglandin synthesis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-13301