Learn More
Apexbio Technology LLC Amitriptyline HCl 549-18-8 1g
SDP
Supplier:  Apexbio Technology LLC B22311000
Encompass_Preferred
Amitriptyline HCl (CAS 549-18-8) is a tricyclic compound that modulates multiple neurotransmitter systems It acts as an inhibitor at serotonin and norepinephrine receptors demonstrating potent activity with IC50 values of 3 45 nM and 13 3 nM respectively Amitriptyline also exhibits notable affinity for 5-HT4 (IC50 7 31 nM) 5-HT2 (IC50 235 nM) and sigma-1 receptors (IC50 287 nM) indicating a broad spectrum of receptor interactions This pharmacological profile supports its application in neuropharmacological research including studies of synaptic transmission antidepressant mechanisms and receptor signaling pathways
Catalog No. 50-224-0419