Learn More
Selleck Chemical LLC Amitriptyline HCl S3183-1g
SDP
Supplier:  Selleck Chemical LLC S31831G
Encompass_Preferred
Amitriptyline HCl is an inhibitor of both serotonin transporter (SERT) and norepinephrine transporter (NET) with Ki of 3 45 nM and 13 3 nM respectively Amitriptyline HCl also inhibits histamine receptor H1 histamine receptor H4 5-HT2 and sigma 1 receptor with Ki of 0 5 nM 7 31 nM 235 nM and 287 nM respectively Amitriptyline is a tricyclic antidepressant (TCA)
Catalog No. 50-311-0423