Learn More
Cayman Chemical AmlodIpIn-d4maleate 1mg
SDP
Supplier:  Cayman Chemical 250351 MG
Encompass_Preferred
An internal standard for the quantification of amlodipine by GC- or LC-MS
Catalog No. 50-293-4671