Learn More
Enzo Life Sciences Amlodipine, (100mg), CAS Number: 88150-42-9
SDP
Supplier:  Enzo Life Sciences BMLCA2020100
Encompass_Preferred
Ca2+ channel blocker. Amlodipine is a third generation L-type calcium channel blocker with a unique spectrum of actions, over and above its antihypertensive activity which include stimulation of NO synthase, inhibition of monocyte-endothelial adhesion, inhibition of oxidative stress and sustained duration of action. Formula: C20H25ClN2O5. MW: 408.9. CAS Number: 88150-42-9. Purity: ≥98% (Assay). Appearance: Almost white crystal powder. Solubility: Soluble in methanol, DMSO (25mg/ml), 100% ethanol (25mg/ml), dimethyl formamide (25mg/ml) or chloroform. Long Term Storage: Ambient
Catalog No. NC0694132