Learn More
Selleck Chemical LLC Amlodipine maleate
SDP
Supplier:  Selleck Chemical LLC S596625MG
Encompass_Preferred
Amlodipine (Amvaz UK-48340) maleate is a long-acting calcium channel blocker used to lower blood pressure and prevent chest pain
Catalog No. 50-000-03322