Learn More
Hach Company Ammonia Cyanurate Reagent Powder Pillows,25 mL, pk/50
Supplier:  Hach Company 2395566
Encompass
For determinaton of Ammonia by the Salicylate Method. Hach method 8155. Range: 0.01 - 0.50 mg/L NH3-N. Sample Size: 25 mL. Pack of 50 Powder Pillows.
Catalog No. 50-199-5912
 Restricted Item