Learn More
Aqua Solutions Ammonia Standard APHA 2502 ppm as NH₃ (1ml = 2.5mg NH₃) (4L)
SDP
Supplier:  Aqua Solutions 03744LC
Encompass_Preferred
Ammonia Standard APHA 2502 ppm as NH₃ (1ml = 2.5mg NH₃) (4L)
Catalog No. NC9252774