Learn More
Hach Company Ammonia TNTplus Vial Test, ULR (0.015 - 2.00 mg/L NH₃-N), 25 Tests
Supplier:  Hach Company TNT830
Encompass
Ammonia TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime. Robust ammonia reagents can be used to determine ammonia levels in a variety of samples including drinking water and wastewater, as well as environmental waters and watershed protection monitoring. Hach Method 10205 is a Salicylate based ammonia chemistry and Equivalent to EPA 350.1, EPA 351.1, and EPA 351.2. Simply add sample to the Ammonia reagent vial, invert the cap, and read in your Hach spectrophotometer. No more dealing with traditional mercury based chemistry and large amounts of waste and glassware!
Catalog No. 50-199-8125
 Restricted Item