Learn More
Selleck Chemical LLC Ammonium ferric citrate
SDP
Supplier:  Selleck Chemical LLC E03751G
Encompass_Preferred
Ammonium ferric citrate (Ferric ammonium citrate Ammonium iron(III) citrate Iron ammonium citrate FerriSeltz) induces ferroptosis in non-small-cell lung carcinoma through the inhibition of GPX4-GSS/GSR-GGT axis activity
Catalog No. 50-313-7041