Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium hexafluorophospha1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 216593
Encompass
Ammonium hexafluorophosphate decomposes on heating before melting. Solubility of ammonium hexafluorophosphate in H2O at 20oC is 74.8% w/v. Additionally, it is very soluble in Me2CO, MeOH, EtOH and MeOAc, and is decomposed by boiling mineral acids.
Catalog No. 50-290-6716