Learn More
Medchemexpress LLC Ammonium lactate | 515-98-0 | 60.0% | 25 G

Supplier: Medchemexpress LLC 25GHYB1530
Ammonium lactate is the ammonium salt of lactic acid. It possesses mild anti-bacterial properties and can be utilized in research pertaining to xerosis.
- Mild anti-bacterial properties.
- Suitable for xerosis research.
- Presented as a colorless to light yellow liquid.
- Recommended storage at -20°C for 2 years as a solution.
- Soluble in water at 116.67 mg/mL.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.