Learn More
Ammonium Molybdate, TS for Phosphate, Reagents
Ammonium Molybdate, TS for Phosphate, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.
$114.40 - $259.00
Chemical Identifiers
| CAS | 7732-18-5 , 7697-37-2 , 7697-37-2 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-335-879
|
Reagents Holdings Llc
CA221500120C |
120 mL |
Each for $114.40
|
|
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
01-335-878
|
Reagents Holdings Llc
CA221500500C |
500 mL |
Each for $259.00
|
|
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.