Learn More
Ammonium Molybdate, TS for Phosphate, Reagents
Specifications
Specifications
| Color | Colorless to Light Yellow |
| Grade | USP |
| Packaging | Glass Bottle |
| Specific Gravity | 1.7 |
| Quantity | 120 mL |
| Sterility | Non-sterile |
| Physical Form | Liquid |
| Chemical Name or Material | Ammonium Molybdate, TS for Phosphate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.