Learn More
Medchemexpress LLC Ammonium polyphosphate, n<20, water solubility>90 g/100 mL | 68333-79-9 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY109536B
Encompass_Preferred
Ammonium polyphosphate, n<20, water solubility>90 g/100 mL is a low-polymerization-degree ammonium polyphosphate (APP) that serves as an important raw material in chelated and controlled-release fertilizers. This product is intended for research use only and is not sold to patients.
  • Low-polymerization-degree ammonium polyphosphate (APP).
  • Important raw material in chelated fertilizers.
  • Important raw material in controlled-release fertilizers.
  • Water solubility >90 g/100 mL.
Catalog No. 50-003-47718