Learn More
Medchemexpress LLC Ammonium trichloro(dioxoethylene-o,o')tellurate | 106566-58-9 | 98.0% | 312.05 | C2H8Cl3NO2Te | 10 MG

Supplier: Medchemexpress LLC HY10101910MG
Ossirene (AS101) is an immunomodulatory tellurium compound used in research as a potent IL-1β inhibitor. It abolishes phosphorylation of STAT3 by inhibiting IL-10 and potently inhibits caspase-1, making it useful for studies of autoimmune disease and certain malignancies.
- Potent IL-1β inhibitor.
- Inhibits caspase-1.
- Abolishes STAT3 phosphorylation via IL-10 inhibition.
- Purity ≥98.0%.
- Solid, white to off-white appearance.
- Molecular weight 312.05 g/mol.
- Supplied as 10 mg.
- Store at -20°C under nitrogen; in solvent, -80°C (up to 6 months).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.